Skip to Content
Merck
All Photos(1)

Key Documents

14714

Sigma-Aldrich

Cesium

purum, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cs
CAS Number:
Molecular Weight:
132.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.5%

vapor pressure

1 mmHg ( 279 °C)

grade

purum

Assay

≥99.5%

reaction suitability

reagent type: reductant

resistivity

19 μΩ-cm, 0°C

bp

705 °C (lit.)

mp

28.5 °C (lit.)

density

1.873 g/mL at 25 °C (lit.)

SMILES string

[Cs]

InChI

1S/Cs

InChI key

TVFDJXOCXUVLDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Investigations studying scattering effects on quantum efficiency and response time for cesiated metal photocathodes

Alkali modifier for hydrogenation catalysts

Reactant involved in synthesis of pseudo-hollandite chromium tellurides

Catalyst for ammonia synthesis, investigations of in situ cesium reduction

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ilenia Rossetti et al.
Inorganic chemistry, 50(8), 3757-3765 (2011-03-26)
We present here a X-ray absorption spectroscopy (XAS) investigation on the local chemical order and electronic structure of Cs and Ba, promoters of the Ru/C catalysts for ammonia synthesis that attracted interest because of highly increased productivity. The role of
S. Yamazaki and Y. Ueda,
Diffusion and Defect Data, Solid State Data, Part B, 170, 17-20 (2011)
R. Kosydar, et al.,
Applied Catalysis, 402, 121-131 (2011)
K. L. Jensen, et al.,
Journal of Applied Physics, 110 (2011)
Ying Wang et al.
Chemical communications (Cambridge, England), 49(3), 306-308 (2012-11-28)
A noncentrosymmetric chiral Strandberg-type polyoxometalate based material, Cs(4)Mo(5)P(2)O(22), that contains unusual one dimensional chains constructed from [Mo(5)P(2)O(23)](6-) units has been reported. Cs(4)Mo(5)P(2)O(22) shows a moderate SHG response, and is phase-matchable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service