Skip to Content
Merck
All Photos(1)

Key Documents

OP66

Sigma-Aldrich

Anti-Rb (Ab-5) Mouse mAb (LM95.1)

liquid, clone LM95.1, Calbiochem®

Synonym(s):

Anti-Retinoblastoma Associated Protein, Anti-pp105

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
clone:
LM95.1, monoclonal
application:
species reactivity:
human
citations:
17

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

LM95.1, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RB1(5925)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells. Recognizes the ~105 kDa Rb protein.
Recognizes the ~105 kDa Rb protein. Sold under license of U.S. Patent 5,853,988 and corresponding patents.
This Anti-Rb (Ab-5) Mouse mAb (LM95.1) is validated for use in Immunoblotting, Paraffin Sections for the detection of Rb (Ab-5).

Immunogen

Epitope: within a 56 kDa C-terminal fragment
Human
recombinant, human Rb

Application

Immunoblotting (1 µg/ml; see application references)

Paraffin Sections (3 µg/ml, see comments and application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Negative Control
Saos cells
Positive Control
ML-1 cells or normal colon tissue

Other Notes

Dyson, N., et al. 1992. J. Virol.66, 4606.
Dyson, N., et al. 1990. J. Virol.64, 1353.
Horowitz, J.M., et al. 1989. Science243, 937.
Whyte, P., et al. 1988. Nature334, 124.
Klein, G. 1987. Science238, 1539.
Lee, W.H., et al. 1987. Science235, 1394.
Lee, W.H., et al. 1987. Nature329, 642.
Knudson, A.G. 1971. Proc. Natl. Acad. Sci. USA68, 820.
Pre-treatment with heat or pressure cooker may improve staining of some tissue specimens; this should be determined for each specimen. Antibody should be titrated for optimal results in individual systems.

Legal Information

Sold under license of U.S. Patent 5,853,988 and corresponding patents.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abstracts from USCAP 2019: Genitourinary Pathology (including renal tumors) (776-992).
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 32(Suppl 2), 1-173 (2019-03-20)
Alessandro Bertolo et al.
Scientific reports, 10(1), 19084-19084 (2020-11-07)
Mesenchymal stromal cells (MSC) are used in cell therapy, but results depend on the unknown quality of cell populations. Extended culture time of MSC increases their senescent levels, leading to a critical loss of cell fitness. Here, we tested the
Ching-Wen Tsai et al.
Artificial cells, nanomedicine, and biotechnology, 46(8), 1852-1863 (2017-10-31)
This study evaluated the effect of chitosan, poly vinyl alcohol (PVA) and poly (2-hydroxyethyl methacrylate) (pHEMA) on delaying the human fibroblast senescence. Cells could form suspending multicellular spheroids on these biomaterials, but only chitosan was capable of decreasing the SA
Tao Wei et al.
mBio, 12(4), e0227721-e0227721 (2021-09-02)
The species specificity of papillomaviruses has been a significant roadblock for performing in vivo pathogenesis studies in common model organisms. The Mus musculus papillomavirus type 1 (MmuPV1) causes cutaneous papillomas that can progress to squamous cell carcinomas in laboratory mice.
Hsi-Hsien Hsieh et al.
Journal of cell science, 130(19), 3272-3281 (2017-08-11)
SerpinB2 is a serine protease inhibitor also known as plasminogen activator inhibitor type 2 (PAI-2). It has been well documented that serpinB2 is an inhibitor of urokinase plasminogen activator (uPA) and tissue plasminogen activator (tPA). Interestingly, serpinB2 levels are increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service