Principaux documents
55071
Adaptateur de tube
1/4-28 female (flat-bottom), 10-32 male (standard)
About This Item
Code UNSPSC :
41115700
Matériaux
PEEK adapter (one-piece design)
Description
for chromatography applications
Conditionnement
pkg of 1 ea
Diam. de l'orifice
0.030 in.
Raccords
1/4-28 female (flat-bottom)
10-32 male (standard)
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis