Principaux documents
Z757373
Sartorius Proline® Plus mechanical pippettor
single channel, volume 10-100 μL
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Caractéristiques
electronic: no
single-channel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Sartorius 728050
Volume
10-100 μL
Description générale
The Sartorius Proline Plus pipettes of the Biohitfamily are high quality with a light pipetting force. With robust design, the Proline Plus is suitable for variable every day laboratory applications and covers the full range of 0.1μL to 10 mL.
  • Reduced pipetting forces prevent RSI and improve the results
  • Accurate and precise
  • Contamination free pipetting with Safe-Cone Filters
  • Robust design
  • Easiest pipette to maintain
Informations légales
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy