Principaux documents
Z664650
Brady® Freezerbondz labels
L × W 31.75 mm × 9.53 mm
About This Item
Code UNSPSC :
55121611
Matériaux
polyester
white
Description
for LabXpert printer only
Conditionnement
roll of 450 labels
Fabricant/nom de marque
Brady X-112-490
Technique(s)
cryopreservation: suitable
L × l
31.75 mm × 9.53 mm
Description générale
Thermal transfer printable labels capable of adhering to cold/frozen surfaces. Constructed from white polyester.
  • Thermal transfer printing offers crisp, high resolution, smear-free print
  • Can be used to indentify lab samples that are already frozen
  • Can survive environments of down to -196°C for a minimum of ten years
  • Excellent resistance to laboratory chemicals
  • Available in sizes to fit most popular tubes, vials, conicals or trays
Informations légales
Brady is a registered trademark of Brady Worldwide, Inc.
FreezerBondz is a trademark of Brady Worldwide, Inc.