Principaux documents
Z640166
Eppendorf® epT.I.P.S. Biopur®, singles
volume 0.1-20 μL
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.02
Longueur:
40 mm
Volume maximum:
20 μL
Stérilité:
sterile
Conditionnement:
pack of 100 ea (individually wrapped)
Fabricant/nom de marque:
Eppendorf® 0030 010.019
Eppendorf® 022491130
Matériaux
colorless
Stérilité
sterile
Conditionnement
pack of 100 ea (individually wrapped)
Fabricant/nom de marque
Eppendorf® 0030 010.019
Eppendorf® 022491130
Plage de mesure du volume
0.1-20 μL
Longueur
40 mm
Volume maximum
20 μL
Description générale
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Informations légales
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG