Principaux documents
Z380296
Jars Nalgene® style 2117
capacity 125 mL
Synonyme(s) :
laboratory jars, nalgene jars
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
polymethylpentene bottle
polypropylene cap
polypropylene screw closure
round transparent bottle
Joint
Joints threaded neck (70)
Description
Autoclavable with closure completely unscrewed
Caractéristiques
with cap
Conditionnement
pkg of 4
Fabricant/nom de marque
Nalgene® 2117-0125
Diam. du flacon × H
64 mm × 74 mm
Capacité
125 mL
Description générale
Transparent, with good chemical resistance.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries