Z340553
BRAND® BISTABIL® ground glass stopper, conical joint
joint: ST/NS 12/21
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.35
Produits recommandés
Joint
joint: ST/NS12/21
Fabricant/nom de marque
BRAND 144615
Description générale
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Autres remarques
BR144615 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences