Principaux documents
RFA452411K-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-24-11-Kit (rotor with higher edge), aerosol-tight, to hold, 24 x spin columns, lid
About This Item
Code UNSPSC :
41103900
Caractéristiques
lid
to hold 24 x spin columns
aerosol-tight
fixed angle rotor
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022654128 (NA)
Eppendorf® 5427752005 (EU)
Paramètres
19,090 × g max. RCF
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5430/5430R
Description générale
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG