Principaux documents
OH30378542
Ohaus® Dissolved Oxygen Meter
Model ST400D-G
About This Item
Code UNSPSC :
41115611
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30378542
dissolved oxygen measuring range
0.00-20.0 mg/L (0 - 50 °C)
Alimentation par piles
4 AAA, 250 hr life
Description générale
Large backlit LCD screen provides easy to read results - even in low light environments. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
Application
Field and lab Dissolved Oxygen measurement
Caractéristiques et avantages
99 measurement memory, last calibration data recall, auto barometric pressure measurement, auto temperature compensation with manual salinity compensation
Informations légales
Ohaus is a registered trademark of Ohaus Corp.