Principaux documents
OH30219114
Ohaus® pH Meter
Model ST300-G
About This Item
Code UNSPSC :
41115603
Matériaux
epoxy shaft
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30219114
ORP measuring range
-1999-+1999 mV
Plage de mesure du pH
0.00-14.00 pH unit
Plage de mesure de la température
0-100 °C
Alimentation par piles
4 AAA, 250 hr life
Description générale
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
Application
pH, oxidation-reduction potential (ORP) measurement
Caractéristiques et avantages
Automatic and manual temperature compensation, auto buffer recognition, calibration icon
Informations légales
Ohaus is a registered trademark of Ohaus Corp.