Principaux documents
OH30058731
Ohaus® Conductivity Meter
Model ST3100C-F, AC/DC input 120 V, US 3-pin plug
About This Item
Code UNSPSC :
41115609
Entrée CA/CC
110/120 V, US/Japan plug
Matériaux
epoxy shaft
Caractéristiques
CSA approved
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30058731
Plage de mesure de la conductivité
0.00-199.9 mS/cm
Plage de mesure de la température
0-100 °C
Description générale
Among Starter 3100C′s user-friendly features that support ease-of-use are an adjustable standalone electrode holder and large backlit LCD display.
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results.
Application
Conductivity, total dissolved solids (TDS) measurement
Conductivity, total dissolved solids (TDS), salinity measurement
Caractéristiques et avantages
99 measurement memory, automatic and manual endpoint functions, automatic and manual temperature compensation
Automatic temperature compensation, adjustable temperature coefficient
Informations légales
Ohaus is a registered trademark of Ohaus Corp.