Principaux documents
OH30038553
Ohaus® ORP Electrode, STORP1
STORP1
About This Item
Code UNSPSC :
39121436
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30038553
ORP measuring range
-1000-+1000 mV
Description générale
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Caractéristiques et avantages
Can be used in conjunction with all Starter bench and portable meters
Informations légales
Ohaus is a registered trademark of Ohaus Corp.