Principaux documents
EP5804770005
Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL
holds 4 tubes 1.5-2 mL, pack of 2 ea
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.82
Caractéristiques
holds 4 tubes 1.5-2 mL
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Eppendorf® 5804770005
Technique(s)
centrifugation: suitable
Compatibilité
for use with Rotor F-34-6-38
Informations légales
Eppendorf is a registered trademark of Eppendorf AG