Principaux documents
EP5391070018
Eppendorf(R) Plate Adapter HeatSealer S100
for 96 and 384 well plates, low profile
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.15
Caractéristiques
for 96 and 384 well plates
low profile
Fabricant/nom de marque
Eppendorf® 5391070018 (Global)
Catégories apparentées
Caractéristiques et avantages
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Informations légales
Eppendorf is a registered trademark of Eppendorf AG