Principaux documents
EP2231302009
Eppendorf®Reference 2 Variable Volume Pipettor Kits
no. of channels, Single Channel, pack of 6, volume 0.5—10 μL, volume 10—100 μL, volume 30—300 μL, volume 100—1,000 μL, volume 0.5—5 mL, volume 1—10 mL, includes tips
Synonyme(s) :
Racks
About This Item
Code UNSPSC :
41121500
Caractéristiques
adjustable
manual
no. of channels Single Channel
Conditionnement
pack of 6
Fabricant/nom de marque
Eppendorf® 2231302009
Volume
0.5—10 μL
0.5—5 mL
100—1,000 μL
10—100 μL
1—10 mL
30—300 μL
Catégories apparentées
Description générale
Eppendorf® Reference 2, 6-pack Option 2, 1-channel, variable, includes 6 adjustable-volume pipettes (0.5-10 µL, 10-100 µL, 30-300 µL, 100-1000 µL, 0.5-5 mL, 1-10 mL), 1 full box of Eppendorf pipette tips for each pipette volume (excludes 5 mL and 10 mL tips), 0.1 µL – 10 mL
Caractéristiques et avantages
  • Mechanical air-cushion pipette with outstanding precision and accuracy - operated with just one button for volume selection, pipetting and tip ejection
  • With its innovative one-button operation, the Reference 2 allows fast and ergonomic handling with reduced operating efforts and active aerosol reduction.
  • A clear haptic feedback helps to avoid accidental tip ejection 4-digit display with magnifying window for quick and secure volume setting and identification (clearly visible from every angle), incl. volume lock
  • Spring-loaded tip cones improve user-to-user reproducibility and ergonomics by lowering the tip attachment force
  • The unique smooth surface and full autoclavability guarantee efficient decontamination - making it the ideal companion when working under sterile conditions
  • A temporary adjustment allows for an easy adjustment of your Reference 2 for the most accurate pipetting of different liquids or in changing external conditions without the need for a full calibration
  • Available as single-channel pipette in fixed or variable volume as well as 8- and 12-channel pipette
  • The Reference 2 multi-channel variants offer high flexibility incl. a spring-loaded tip cone which can be switched on/off optionally
Informations légales
Eppendorf is a registered trademark of Eppendorf AG