EP022822101
Rotor Spacer
required for stacking rotors Eppendorf® F-45-72-8 and F-45-48-11 rotors
About This Item
Produits recommandés
Caractéristiques
required for stacking rotors Eppendorf® F-45-72-8 and F-45-48-11 rotors
Fabricant/nom de marque
Eppendorf® 022822101
Technique(s)
centrifugation: suitable