CLS48012
Corning® LSE™ Digital Vortexer Spring Loaded Platfrom for Flasks, Bottles, or Tube Racks
Synonyme(s) :
Dnase and Rnase free flask, Erlenmeyer flask, conical flasks, flasks
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.61
Produits recommandés
Conditionnement
case of 1 ea
pkg of 1 pkg
Fabricant/nom de marque
Corning 48012
48012
Catégories apparentées
Description générale
This is one of three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes.
Informations légales
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.