AXYPCR384M2Y
PCR microplate for ABI 3730, 3100 and MJ Thermocyclers
size 384 wells, conical bottom yellow polypropylene wells, pkg of 5x10plates/cs
Synonyme(s) :
384 multiwell plate
About This Item
Matériaux
conical bottom yellow polypropylene wells
Stérilité
non-sterile
Fabricant/nom de marque
Axygen Inc PCR-384M2-Y
Corning
Conditionnement
pkg of 5x10plates/cs
Volume maximum
50 μL
Taille
384 wells
Volume de travail
50 μL
Type de liaison
non-treated surface