Key Documents
Z801135
Hellma® aluminum spacer
38mmx12.5mmx9 mm, to fit cells with 1 mm light path into 10 mm cell holder
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Fabricant/nom de marque
Hellma 13.101
Technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
Informations légales
Hellma is a registered trademark of Hellma GmbH