Key Documents
Z742667
2MAG steriMIXcontrol for use with steriMIXdrive
AC/DC input 100 V / 240 V AC, US 3-pin plug
Synonyme(s) :
control unit, power bank, power unit
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
Matériaux
stainless steel
Caractéristiques
20 W Stirring Power
Conditionnement
pack of 1 ea
Fabricant/nom de marque
2mag 95200
Paramètres
0-40 °C temp. limit (at 80% humidity)
100-1200 rpm speed
Larg. × P × H
200 mm × 175 mm × 48 mm
Poids
1.8 kg