Z742591
Accuris™ SmartDoc™
2.0 Imaging System with blue light illumination base, AC/DC input 115 V AC, US 3-pin plug
Synonyme(s) :
gel documentation equipment, transilluminator system
About This Item
Produits recommandés
Caractéristiques
12V DC power supply included
High Intensity Blue LED; Orange PMMA photo filter and view cover
includes gel prep/cutting platform, blue light illumination base, orange filter cover, imaging enclosure and orange photo filter
Conditionnement
pkg of 1
Fabricant/nom de marque
Benchmark Scientific E5001-SDB
Larg. × P × H
23 cm × 19 cm × 22 cm, 2.6lbs (1.2kg) weight
Compatibilité
smartphones and tablets with camera
Description générale
Features:
- Ideal for common safe stains (SmartGlow, SYBR Green, GelGreen, etc)
- Visible blue light will not damage DNA
- Quality gel imaging with any smart phone
- Gel access for DNA band cutting
- Imaging enclosure can also be used on existing trans illuminators
- Maximum gel size: 15X15 cm
Les clients ont également consulté
Global Trade Item Number
Référence | GTIN |
---|---|
Z742591-1EA | 4061826162026 |