Z685194
Accessory kit for Crucible Cradle™ and safety tripod stands
for crucibles 5-40 mL
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Produits recommandés
Description générale
Includes:
- (3) locking ceramic triangles for crucibles 5-40 mL
- Locking holder for evaporating dish
- Locking wire gauze pad
Informations légales
Crucible Cradle is a trademark of Global Market Connections, Ltd.