Principaux documents
Z322210
Quickfit® right-angle tubing adapter
joint: ST/NS 34/35
Synonyme(s) :
MF10/5B/SC
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS34/35
Fabricant/nom de marque
Quickfit MF 10/5B/SC
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
With joint and right angle-connection with screw-thread connector for flexible tubing.
Informations légales
Quickfit is a registered trademark of SciLabware Limited