Principaux documents
Z317608
Duran® vacuum desiccator plate
metal plate, I.D. 100 mm
Synonyme(s) :
290804606
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux:
metal plate
D.I.:
100 mm
Matériaux
metal plate
Fabricant/nom de marque
DURAN®
Schott
Disponibilité
not available in USA and Canada
D.I.
100 mm
Description générale
Perforated plate for vacuum desiccators.
Caractéristiques et avantages
• Complies with DIN EN 10143• Manufactured from stainless steel (1.4301, type 304)• The heat-resistant material allows hot samples to be placed into the desiccator• Rust-free
Informations légales
DURAN is a registered trademark of DWK Life Sciences