Key Documents
Z303313
Quickfit® jacketed coil condenser
L 160 mm, joint: ST/NS 24/29
Synonyme(s) :
CX6/33/SC
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/29
Fabricant/nom de marque
Quickfit CX 6/33/SC
Disponibilité
not available in USA and Canada
not available in USA and Canada
Longueur
160 mm
Superficie
400 cm2
Description générale
Double surface condensers with a coil as the inner surface provides approximately twice the surface area of plain, double surface condensers of the same effective length.
Informations légales
Quickfit is a registered trademark of SciLabware Limited