Principaux documents
Z245208
Sigma-Aldrich
Bransonic® Model B200 cleaner
AC/DC input 115 V AC
Synonyme(s) :
ultrasonic bath sonicator, ultrasonic cleaning bath
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.74
Caractéristiques
CSA approved
CSA approved
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Branson 100-951-010
Disponibilité
not available in EU
Description générale
Low-cost, portable, ultrasonic cleaner has the same cleaning ability as larger models. SS tank with Noryl® base, parts basket, and plastic cover. Cleaner automatically shuts off after 5 min.
Informations légales
Bransonic is a registered trademark of Branson Ultrasonics Corp.
Noryl is a registered trademark of Sabic Innovative Plastics IP B.V.
Global Trade Item Number
RéférenceGTIN
Z245208-1EA4061833293157