Principaux documents
Z174955
Conical reaction vial
capacity 3 mL
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Ace Glass 9591-16
Capacité
3 mL
Description générale
Thin-wall with threaded outer 14/10 joint, cap with hole, PTFE-faced silicone rubber septum, and FETFE® O-ring.
The product does NOT have the side arm opening. It is the product shown on the left of the photo.
Informations légales
FETFE is a registered trademark of Ace Glass, Inc.
Global Trade Item Number
RéférenceGTIN
Z174955-1EA4061832348131