Z135631
Burette clamp
single
Synonyme(s) :
burette clamps, clamp lab, lab clamp
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Description générale
Noncorroding HDPE. Concealed steel spring and rubber pads prevent slipping. Center clamp tightens to fit standard support rod; 1/2 and 3/8 in. bushings included.