Key Documents
THS01435
ThalesNano CatCart® catalyst cartridge system, 70 mm L
1% Pt/SiO2/polyethyleneimine
Synonyme(s) :
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Code UNSPSC :
41103817
Nomenclature NACRES :
NB.51
Conditionnement
pack of 1 ea
Fabricant/nom de marque
ThalesNano THS 01435
Diamètre
4 mm
Description générale
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Informations légales
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.