Principaux documents
TEM-Q250AR1.3
Sigma-Aldrich
QUANTIFOIL R 1.2/1.3
Gold Grids, size 200 mesh, pack of 50
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Taille:
200 mesh
Conditionnement:
pack of 50
Conditionnement
pack of 50
Taille
200 mesh
Description générale
QUANTIFOIL with circular holes is used in cryoelectron tomographic reconstruction. The roundness of the holesis advantageous with respect to the formation of an icelayer of constant thickness. Assessment of the image quality is easier when foil is incl