Principaux documents
TEM-G400F1-CU
Sigma-Aldrich
Gilder Finder Grids G400F1
Copper, size 400 mesh, vial of 100
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Taille:
400 mesh
Conditionnement:
vial of 100
Conditionnement
vial of 100
Taille
400 mesh
Description générale
Gilder Finder Grids G400F1. Copper grids, 400 mesh. The block of 1152 (32x36) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 36 (6x6) cells has a central number from 1 to 32 for identification purposes.