Principaux documents
TEM-FF150CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 150 mesh, box of 50
Synonyme(s) :
FORMVAR SQUARE MESH - CU 150 MESH UL 50/BX
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 50
Taille
150 mesh
Description générale
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 150 mesh. All of our film is laid on the shiny side of the grid.