Principaux documents
SYNF762450
Synthware storage tube with standard taper side arm
capacity 50 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonyme(s) :
Synthware F762450, storage tube with outer joint
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
black cap
cap (plug seal)
colorless tube
glass tube
round bottom
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F762450
Capacité
50 mL
Taille de la vanne
0-4 mm
Description générale
Supplied with high vacuum valve with PTFE O-ring at tip.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Global Trade Item Number
RéférenceGTIN
SYNF762450-1EA4061832196121