Principaux documents
SYNC111420
Synthware cold finger condenser
joint: ST/NS 14/20, hose size 8 mm
Synonyme(s) :
Synthware C111420, Cold finger condenser with drip tip
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C111420
Taille du tuyau
8 mm
Description générale
Extended offset lower tip for drop control.
Informations légales
Synthware is a trademark of Kemtech America Inc.