Key Documents
SYNA272414
Synthware anti-climb adapter
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 14/20
Synonyme(s) :
Synthware A272414, splash-guard adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS14/20
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A272414
Capacité
100 mL
Description générale
Trap provides expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the The inner vapor tube is sealed to the top of thetrap to prevent entry into the condenser.
Informations légales
Synthware is a trademark of Kemtech America Inc.