Principaux documents
DWK241935702
DURAN® Erlenmeyer flask with standard ground joint
flask capacity 1,000 mL, neck joint: ST/NS 45/40
Synonyme(s) :
241935702
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass 3.3 flask
flat
Joint
joint: ST/NS neck45/40
Mode de stérilisation compatible
autoclavable compatible
Caractéristiques
without cap
Conditionnement
case of 10 ea
Fabricant/nom de marque
DURAN® 241935702
Contenance du flacon/de la fiole
1,000 mL
Description générale
Tight closure due to standard ground joint. Fired-on, highly durable white print. Easy mixing due to conical form
Autres remarques
Informations légales
DURAN is a registered trademark of DWK Life Sciences