Principaux documents
CLS31770500EC
Pyrex® gas-washing bottle, fritted cylinder, 29/42 ST stopper
capacity 500 mL
Synonyme(s) :
Corning® 31770
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass bottle
clear bottle
cylindrical bottle
glass cap
straight bottle
Caractéristiques
cap
fritted
Conditionnement
case of 1 ea
pack of 1 ea
Fabricant/nom de marque
Corning 31770-500EC
Capacité
500 mL
Diam. × H
63 mm × 290 mm
Dimension de pores
170-220 μm porosity
40-60 μm porosity (coarse)
coarse (extra)
Description générale
12mm diameter fritted cylinder.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.