Principaux documents
CLS2480600
Pyrex® Allihn condenser, with male and female joints
jacket L 600 mm, joint: ST/NS 29/42
Synonyme(s) :
laboratory condenser
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS29/42
Fabricant/nom de marque
Corning 2480-600
L de l'enveloppe
600 mm
Catégories apparentées
Description générale
Suitable for use in vacuum distillation. With drip-tip
Informations légales
PYREX is a registered trademark of Corning, Inc.