Principaux documents
CLS2480200
Pyrex® Allihn condenser, with male and female joints
jacket L 200 mm, joint: ST/NS 19/38
Synonyme(s) :
laboratory condenser
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS19/38
Fabricant/nom de marque
Corning 2480-200
L de l'enveloppe
200 mm
Description générale
Suitable for use in vacuum distillation. With drip-tip
Informations légales
PYREX is a registered trademark of Corning, Inc.