Principaux documents
BR704548
BRAND® replacement pump assembly for seripettor® pro
size 10 mL
Synonyme(s) :
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
Code UNSPSC :
40151500
Nomenclature NACRES :
NB.01
Matériaux
(Polyphenylene Oxide/polyetherimide (UV protection))
(Hastalloy (stainless))
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 704548
Taille
10 mL
Description générale
The BRAND® Replacement Pump Assembly for the Seripettor® Pro is designed for precise and efficient liquid handling in laboratory applications. This actuating unit is constructed from high-quality materials, including polyphenylene oxide (PPO) and polyetherimide (PEI) for UV protection, ensuring durability and longevity. The assembly features a Hastelloy® lifting spring, which enhances performance and reliability. This replacement pump assembly is essential for maintaining the functionality of your Seripettor® Pro, making it an indispensable tool for researchers and technicians seeking accurate and efficient liquid dispensing.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Hastelloy is a registered trademark of Haynes International, Inc.
seripettor is a registered trademark of BRAND GMBH + CO KG