Merck
Toutes les photos(6)
Principaux documents
BAF371220012
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated, L × diam. 0.4375 in. (12 mm) × 0.1875 in. (4.5 mm)
Synonyme(s) :
Spinbar® magnetic stir bar
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
Matériaux
polygon white PTFE-coated
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F37122-0012
L × diam.
0.4375 in. (12 mm) × 0.1875 in. (4.5 mm)
Description générale
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
Caractéristiques et avantages
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company