Skip to Content
Merck
All Photos(1)

Key Documents

T0781

Supelco

3,5,5,-Trimethyloxazolidine-2,4-dione

analytical standard

Synonym(s):

Trimethadione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO3
CAS Number:
Molecular Weight:
143.14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CN1C(=O)OC(C)(C)C1=O

InChI

1S/C6H9NO3/c1-6(2)4(8)7(3)5(9)10-6/h1-3H3

InChI key

IRYJRGCIQBGHIV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tammye L Fleeman et al.
Birth defects research. Part B, Developmental and reproductive toxicology, 71(3), 185-190 (2004-07-30)
Congenital membranous ventricular septal defects (VSD) have been shown to close during postnatal development in rats [Solomon et al., Teratology 55:185-194, 1997]. Although they may differ in size, spontaneous and treatment-related VSD are histologically similar; however, the postnatal fate of
Education and imaging. Hepatobiliary and pancreatic: oral therapy for pancreatic duct stones.
A Hashimoto et al.
Journal of gastroenterology and hepatology, 21(9), 1496-1496 (2006-08-17)
H Nakamura et al.
Journal of clinical pharmacy and therapeutics, 26(6), 417-424 (2001-11-28)
The trimethadione (TMO) tolerance test was performed to evaluate its usefulness in the assessment of hepatic functional reserve in patients with biliary atresia. Nineteen patients with biliary atresia after hepatic portoenterostomy (age range: 2 months to 25 years; sex: 6
Haiyan Shen et al.
Hearing research, 226(1-2), 52-60 (2007-02-13)
Cochlear noise injury is the second most frequent cause of sensorineural hearing loss, after aging. Because calcium dysregulation is a widely recognized contributor to noise injury, we examined the potential of calcium channel blockers to reduce noise-induced hearing loss (NIHL)
N Kurata et al.
Xenobiotica; the fate of foreign compounds in biological systems, 28(11), 1041-1047 (1999-01-08)
1. Caucasian liver samples were used in this study. N-demethylation of trimethadione (TMO) to dimethadione (DMO) was monitored in the presence of chemical inhibitors of CYPs, such as fluconazole, quinidine, dimethyl-nitrosamine, acetaminophen, phenacetin, chlorzoxazone and mephenytoin. Trimethadione N-demethylation was selectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service