Skip to Content
Merck
All Photos(1)

Key Documents

ST1672

Sigma-Aldrich

Anti-DPYD Mouse mAb (7D4)

liquid, clone 7D4, Calbiochem®

Synonym(s):

Anti-Dihydropyrimidine Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7D4, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPYD(1806)

General description

Anti-DPYD, mouse monoclonal, clone 7D4, recognizes the ~100 kDa DPYD protein in HeLa and HeLa NE cells. It is validated for use in ELISA, Western blotting, and immunocytochemistry.
Purified mouse monoclonal antibody. Recognizes the ~100 kDa DPYD protein.
Recognizes the ~100 kDa DPYD protein in HeLa and HeLa NE cells.

Immunogen

A recombinant polypeptide corresponding to amino acids of 1-111 human DPYD, expressed as a GST fusion protein

Application

ELISA (see comments)

Immunoblotting (1-5 µg/ml)

Immunocytochemistry (10 µg/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HeLa cells, HeLa NE cells

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service