Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Neutral Red
Synonym(s):
3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Basic Red 5, Toluylene red, Neutral red (C.I. 50040)
Empirical Formula (Hill Notation):
C
15
H
17
ClN
4
CAS Number:
553-24-2
Molecular Weight:
288.78
EC Number:
209-035-8
Products
Product Category
dyes (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
N7005
Neutral Red,
Dye content ≥90 %
Expand
View Pricing
N4638
Neutral Red,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
72210
Neutral Red,
for microscopy (Bact., Bot., Fl., Hist., Vit.), indicator (pH 6.8-8.0)
Expand
View Pricing
1.01369
Neutral Red,
indicator and for microbiology
Expand
View Pricing
861251
Neutral Red,
certified by the Biological Stain Commission
Expand
View Pricing