Skip to Content
Merck
All Photos(1)

Key Documents

V800183

Sigma-Aldrich

Iron(III) nitrate nonahydrate

LR, ≥98%

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystals

grade

LR

product line

Vetec

Assay

≥98%

form

crystals

mp

47 °C (lit.)

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Basler et al.
Proteomics, 6(23), 6194-6206 (2006-11-30)
Restricting bacterial growth by iron-chelating proteins that reduce iron availability in mucosal secretions and body fluids belongs to basic mechanisms of innate immunity. Most pathogens and commensals thus developed gene regulons responding to iron concentration and encoding iron acquisition systems
F Ndong et al.
International endodontic journal, 45(6), 557-564 (2012-01-25)
To determine the effect of increasing the proportion of zinc on partially stabilized cement (PSC) produced using a one-step sol gel process. A one-step sol-gel process of Portland cement-based PSC with Zn was synthesized by replacing iron nitrate. The crystalline
A Roberts et al.
Oral microbiology and immunology, 20(3), 147-153 (2005-04-20)
Psychological stress is known to increase the circulating levels of the catecholamine hormones noradrenaline and adrenaline, which have been shown to influence the growth of a large number of bacterial species by acting in a siderophore-like manner or by inducing
Xianmao Lu et al.
Nano letters, 7(6), 1764-1769 (2007-05-11)
This paper describes a two-step procedure for generating cubic nanocages and nanoframes. In the first step, Au/Ag alloy nanoboxes were synthesized through the galvanic replacement reaction between Ag nanocubes and an aqueous HAuCl4 solution. The second step involved the selective
C L Hsueh et al.
Chemosphere, 58(10), 1409-1414 (2005-02-03)
This study investigated Fenton and Fenton-like reactions at low iron concentration (<or==10 mg l-1) to oxidize three commercial azo dyes, namely Red MX-5B, Reactive Black 5 and Orange G. In some local cases in Taiwan, wastewater color was found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service