Skip to Content
Merck
All Photos(1)

Key Documents

44895-U

Supelco

ASTM® Triolein Solution

5000 μg/mL in pyridine, analytical standard

Synonym(s):

Glyceryl trioleate solution, Biodiesel impurity solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Agency

ASTM® D6584

form

liquid

packaging

pkg of 1 × 2 mL

concentration

5000 μg/mL in pyridine

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

single component solution

InChI

1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-

InChI key

PHYFQTYBJUILEZ-IUPFWZBJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Keller et al.
Neurorehabilitation and neural repair, 26(9), 1080-1088 (2012-05-01)
With progressive abnormalities in leg strength, tone, and sensation, adrenomyeloneuropathy (AMN) is a differential diagnosis for multiple sclerosis and hereditary spastic paraparesis. AMN pathology has been linked to weakness, making it a relevant model to evaluate the relationship between neurodegeneration
Emiko Iwata et al.
Journal of nutritional science and vitaminology, 58(3), 175-180 (2012-08-11)
The albedo is the white part of the citrus peel, which acts as a water reservoir for the juice sacs, seeds and leaves in times of drought. As the functionality of the albedo is unknown, we examined in this study
Colin J MacDougall et al.
Faraday discussions, 158, 425-433 (2012-12-14)
Fats are elastoplastic materials with a defined yield stress and flow behavior and the plasticity of a fat is central to its functionality. This plasticity is given by a complex tribological interplay between a crystalline phase structured as crystalline nanoplatelets
Jonathan C Jun et al.
American journal of physiology. Endocrinology and metabolism, 303(3), E377-E388 (2012-05-25)
Obstructive sleep apnea (OSA) induces intermittent hypoxia (IH) during sleep and is associated with elevated triglycerides (TG). We previously demonstrated that mice exposed to chronic IH develop elevated TG. We now hypothesize that a single exposure to acute hypoxia also
Sylvie Hollebeeck et al.
Food chemistry, 138(2-3), 1936-1944 (2013-02-16)
Bioaccessibility studies should be taken into account when evaluating the physiological effects of ingested compounds at the intestine level. Several in vitro digestion protocols have been described, with a wide range of experimental conditions but no optimised protocol exists. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service