Z664650
Brady® Freezerbondz™ labels
L × W 31.75 mm × 9.53 mm
All Photos(1)
About This Item
Recommended Products
material
polyester
white
description
for LabXpert printer only
packaging
roll of 450 labels
manufacturer/tradename
Brady X-112-490
technique(s)
cryopreservation: suitable
L × W
31.75 mm × 9.53 mm
General description
Thermal transfer printable labels capable of adhering to cold/frozen surfaces. Constructed from white polyester.
- Thermal transfer printing offers crisp, high resolution, smear-free print
- Can be used to indentify lab samples that are already frozen
- Can survive environments of down to -196°C for a minimum of ten years
- Excellent resistance to laboratory chemicals
- Available in sizes to fit most popular tubes, vials, conicals or trays
Legal Information
Brady is a registered trademark of Brady Worldwide, Inc.
FreezerBondz is a trademark of Brady Worldwide, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)