WH0002303M2
Monoclonal Anti-FOXC2 antibody produced in mouse
clone 2H3, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-FKHL14, Anti-MFH1, Anti-forkhead box C2 (MFH-1, mesenchyme forkhead 1)
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2H3, monoclonal
form
buffered aqueous solution
species reactivity
mouse, human, rat
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL
isotype
IgG2bλ
GenBank accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... FOXC2(2303)
Related Categories
General description
Immunogen
Sequence
AASWYLNHSGDLNHLPGHTFAAQQQTFPNVREMFNSHRLGIENSTLGESQVSGNASCQLPYRSTPPLYRHAAPYSYDCTKY
Application
Biochem/physiol Actions
Physical form
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service