SRP2122
CTF1, GST tagged human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Synonym(s):
CTF, CTF/NFI, NF-1, NF-I
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.26
biological source
human
recombinant
expressed in E. coli
Assay
≥70% (SDS-PAGE)
form
frozen liquid
mol wt
~36.8 kDa
packaging
pkg of 10 μg
concentration
650 μg/mL
color
colorless to clear
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human herpesvirus 2 ... CTF-1(1489)
Biochem/physiol Actions
The CCAAT-box binding protein 1 (CTF1, also called NF-I or NF-1) is a proline-rich transcription activator. It selectively activates transcription of genes that contain GCCAAT consensus sequence in the promoter region, including human Ha-Ras, alpha-globin, hsp70 and many other cellular and viral genes. Cloning and functional characterization of the protein revealed that CTF1 contains a N-terminal DNA binding domain and a C-terminal proline-rich activation domain. CTF1 was shown to interact with the general transcription factor IIB (TFIIB) and facilitate the recruitment of TFIIB to TBP-DNA complexes, with the forkhead thyroid transcription factor TTF2 to regulate hormone-dependent transcription of thyroperoxidase gene, as well as other receptor proteins to synergize hormone receptor-responsive activation.
Physical form
Clear and colorless frozen liquid solution
Preparation Note
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N Mermod et al.
Cell, 58(4), 741-753 (1989-08-25)
Human CTF/NF-I consists of a family of CCAAT box binding proteins that activate both transcription and DNA replication. Analysis of cDNA mutants expressed in E. coli and Drosophila cells reveals that the N-terminal portion of CTF-1 is sufficient for site-specific
K A Jones et al.
Cell, 48(1), 79-89 (1987-01-16)
Transcription factor CTF, which is responsible for selective recognition of eukaryotic promoters that contain the sequence CCAAT, was purified to apparent homogeneity by sequence-specific DNA affinity chromatography. Binding sites for CTF in the human Ha-ras and alpha-globin promoters were highly
C Santoro et al.
Nature, 334(6179), 218-224 (1988-07-21)
The CTF/NF-I group of cellular DNA binding proteins recognizes the sequence GCCAAT and is implicated in eukaryotic transcription as well as DNA replication. Molecular analysis of human CTF/NF-I complementary DNA clones reveals multiple messenger RNA species containing alternative coding regions
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service